siRNA Details
| Gene Symbol: KRTCAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173852.3 | NP_776251.1 | 3 | 278 | ||
| GenBank mRNA | AK024167.1 | - | 2 | 363 | ||
| AK311907.1 | - | 3 | 252 | |||
| AY157577.1 | - | 2 | 163 | |||
| BC029806.1 | - | 3 | 265 | 5189880 | ||
| BC048205.1 | - | 3 | 204 | |||
| BC057233.1 | - | 3 | 287 | 6463372 | ||
| BX647396.1 | - | 3 | 207 | |||

KRTCAP2
Genomic Map