siRNA Details
| Gene Symbol: IBA57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010867.3 | NP_001010867.1 | 1 | 303 | ||
| GenBank mRNA | BI820993.1 | - | 1 | 44 | ||
| CN285775.1 | - | 1 | 383 | |||
| DN831582.1 | - | 2 | 249 | |||
| HY081013.1 | - | 1 | 303 | |||

IBA57
Genomic Map