siRNA Details
| Gene Symbol: FBXO45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105573.1 | NP_001099043.1 | 3 | 833 | ||
| GenBank mRNA | AK025697.1 | - | 3 | 561 | ||
| BE778263.1 | - | 3 | 482 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXO45| Gene Symbol: FBXO45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105573.1 | NP_001099043.1 | 3 | 833 | ||
| GenBank mRNA | AK025697.1 | - | 3 | 561 | ||
| BE778263.1 | - | 3 | 482 | |||