siRNA Details
| Gene Symbol: ZNF385C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242704.1 | NP_001229633.1 | 7, 8 | 1030 | ||
| XM_006721751.3 | XP_006721814.1 | 7, 8 | 1138 | |||
| XM_011524479.2 | XP_011522781.1 | 8, 9 | 1834 | |||
| XM_011524482.2 | XP_011522784.1 | 8, 9 | 1831 | |||
| XM_017024314.1 | XP_016879803.1 | 9, 10 | 1963 | |||
| XM_017024315.1 | XP_016879804.1 | 9, 10 | 1798 | |||
| XM_017024316.1 | XP_016879805.1 | 9, 10 | 1960 | |||
| XM_017024320.1 | XP_016879809.1 | 8, 9 | 1225 | |||
| GenBank mRNA | BC080613.1 | - | 3, 4 | 423 | 5922386 | |

ZNF385C
Genomic Map