siRNA Details
| Gene Symbol: STRA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271006.1 | NP_001257935.1 | 4 | 261 | ||
| NM_144998.3 | NP_659435.2 | 3 | 207 | |||
| XM_005256339.2 | XP_005256396.1 | 4 | 413 | |||
| XM_017024326.1 | XP_016879815.1 | 3 | 541 | |||
| XM_017024327.1 | XP_016879816.1 | 4 | 413 | |||
| XM_017024328.1 | XP_016879817.1 | 4 | 282 | |||
| XM_017024329.1 | XP_016879818.1 | 3 | 228 | |||
| GenBank mRNA | BC009571.1 | - | 3 | 176 | 4333630 | |
| BI837753.1 | - | 3 | 175 | |||
| CB110145.1 | - | 4 | 270 | |||
| HY110543.1 | - | 4 | 251 | |||

STRA13
Genomic Map