siRNA Details
| Gene Symbol: SPATA24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194296.1 | NP_919272.1 | 5 | 445 | ||
| XM_005271916.4 | XP_005271973.1 | 5 | 562 | |||
| XM_011543252.2 | XP_011541554.1 | 5 | 446 | |||
| XM_011543253.2 | XP_011541555.1 | 5 | 424 | |||
| GenBank mRNA | AA399058.1 | - | 2 | 228 | ||
| AK098740.1 | - | 5 | 446 | |||
| BC040037.1 | - | 5 | 425 | |||

SPATA24
Genomic Map