siRNA Details
Gene Symbol: SUSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287005.1 | NP_001273934.1 | 6 | 846 | ||
NM_001287006.1 | NP_001273935.1 | 4 | 600 | |||
NM_001287007.1 | NP_001273936.1 | 3 | 411 | |||
NM_001287008.1 | NP_001273937.1 | 4 | 543 | |||
NM_145006.3 | NP_659443.1 | 5 | 732 | |||
XM_011518358.1 | XP_011516660.1 | 8 | 1926 | |||
XM_011518359.1 | XP_011516661.1 | 7 | 1876 | |||
XM_011518361.2 | XP_011516663.1 | 8 | 988 | |||
XM_011518362.1 | XP_011516664.1 | 6 | 1762 | |||
XM_017014448.1 | XP_016869937.1 | 6 | 1033 | |||
XM_017014449.1 | XP_016869938.1 | 5 | 914 | |||
XM_017014450.1 | XP_016869939.1 | 7 | 873 | |||
XM_017014451.1 | XP_016869940.1 | 5 | 1482 | |||
GenBank mRNA | AA490088.1 | - | 2 | 235 | ||
AK128289.1 | - | 3 | 2700 | |||
AL137623.1 | - | 282 | ||||
AY358190.1 | - | 6 | 774 | |||
BC014601.2 | - | 5 | 713 | 4821517 | ||
BC041834.1 | - | 3 | 2626 | 5268318 | ||
BM458621.1 | - | 3 | 289 |
