siRNA Details
| Gene Symbol: VMA21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017980.3 | NP_001017980.1 | 3 | 306 | ||
| XM_011531125.2 | XP_011529427.1 | 3 | 632 | |||
| GenBank mRNA | AK096835.1 | - | 3 | 285 | ||
| AL833596.1 | - | 3 | 259 | |||
| BC103701.1 | - | 3 | 263 | 40023465 | ||
| BC103702.1 | - | 3 | 263 | 40023460 | ||
| BC105693.1 | - | 3 | 263 | 40023462 | ||
| BC105694.1 | - | 3 | 263 | 40023459 | ||
| BC110800.1 | - | 3 | 219 | 6044374 | ||
| DA224893.1 | - | 3 | 306 | |||

VMA21
Genomic Map