siRNA Details
| Gene Symbol: C6orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010868.2 | NP_001010868.2 | 5 | 956 | ||
| XM_005248673.3 | XP_005248730.1 | 4 | 1967 | |||
| XM_006715370.3 | XP_006715433.1 | 4 | 2621 | |||
| XM_006715372.2 | XP_006715435.1 | 2 | 2526 | |||
| XM_011535539.1 | XP_011533841.1 | 3 | 1886 | |||
| XM_017010367.1 | XP_016865856.1 | 4 | 5102 | |||
| GenBank mRNA | AK092941.1 | - | 3 | 1763 | ||
| BC044236.1 | - | 2 | 264 | |||

C6orf163
Genomic Map