siRNA Details
| Gene Symbol: TYSND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040273.2 | NP_001035363.1 | 1 | 1205 | ||
| NM_173555.3 | NP_775826.2 | 1 | 1205 | |||
| GenBank mRNA | BC016840.1 | - | 1 | 659 | ||
| BC030242.1 | - | 1 | 1121 | 5176098 | ||
| BC111501.1 | - | 1 | 1144 | 5732356 | ||

TYSND1
Genomic Map