siRNA Details
| Gene Symbol: OR8G5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005198.1 | NP_001005198.1 | 1 | 500 | ||
| XM_011542662.2 | XP_011540964.1 | 4 | 1054 | |||
| XM_011542663.2 | XP_011540965.1 | 4 | 824 | |||
| XM_011542664.2 | XP_011540966.1 | 3 | 693 | |||
| XM_011542665.2 | XP_011540967.1 | 3 | 974 | |||
| XM_011542666.2 | XP_011540968.1 | 3 | 744 | |||
| XM_011542667.2 | XP_011540969.1 | 2 | 613 | |||
| GenBank mRNA | BC136820.1 | - | 1 | 523 | 9020811 | |
| BC136822.1 | - | 1 | 523 | 9020813 | ||

OR8G5
Genomic Map