siRNA Details
| Gene Symbol: GTSCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278515.1 | NP_001265444.1 | 3 | 468 | ||
| GenBank mRNA | AW082319.1 | - | 1 | 112 | ||
| AY262164.1 | - | 3 | 468 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GTSCR1| Gene Symbol: GTSCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278515.1 | NP_001265444.1 | 3 | 468 | ||
| GenBank mRNA | AW082319.1 | - | 1 | 112 | ||
| AY262164.1 | - | 3 | 468 | |||