siRNA Details
| Gene Symbol: OTUD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145373.2 | NP_001138845.1 | 1 | 1406 | ||
| GenBank mRNA | AB188491.1 | - | 1 | 1406 | ||
| AK096389.1 | - | 1 | 251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTUD1| Gene Symbol: OTUD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145373.2 | NP_001138845.1 | 1 | 1406 | ||
| GenBank mRNA | AB188491.1 | - | 1 | 1406 | ||
| AK096389.1 | - | 1 | 251 | |||