siRNA Details
Gene Symbol: WAC-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033805.1 | 3 | 1114 | |||
GenBank mRNA | AK001143.1 | - | 1 | 325 | ||
BC045818.1 | - | 3 | 1094 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WAC-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033805.1 | 3 | 1114 | |||
GenBank mRNA | AK001143.1 | - | 1 | 325 | ||
BC045818.1 | - | 3 | 1094 |