siRNA Details
| Gene Symbol: PRSS54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080492.1 | NP_001073961.1 | 3, 4 | 469 | ||
| NM_001305173.1 | NP_001292102.1 | 3, 4 | 466 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS54| Gene Symbol: PRSS54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080492.1 | NP_001073961.1 | 3, 4 | 469 | ||
| NM_001305173.1 | NP_001292102.1 | 3, 4 | 466 | |||