siRNA Details
| Gene Symbol: C6orf223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171992.2 | NP_001165463.1 | 2, 3 | 270 | ||
| NM_153246.5 | NP_694978.2 | 2, 3 | 227 | |||
| XM_017010425.1 | XP_016865914.1 | 2, 3 | 1309 | |||
| GenBank mRNA | BC029071.2 | - | 2, 3 | 237 | 4341806 | |
| BC032706.1 | - | 1, 2 | 538 | 5497384 | ||
| BX342526.2 | - | 2, 3 | 254 | |||

C6orf223
Genomic Map