siRNA Details
| Gene Symbol: C6orf223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153246.5 | NP_694978.2 | 1, 2 | 89 | ||
| GenBank mRNA | BC029071.2 | - | 1, 2 | 99 | 4341806 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C6orf223| Gene Symbol: C6orf223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153246.5 | NP_694978.2 | 1, 2 | 89 | ||
| GenBank mRNA | BC029071.2 | - | 1, 2 | 99 | 4341806 | |