siRNA Details
Gene Symbol: LRRC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012974.2 | NP_001012992.1 | 1 | 816 | ||
GenBank mRNA | BC108661.1 | - | 1 | 816 | 5240367 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LRRC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012974.2 | NP_001012992.1 | 1 | 816 | ||
GenBank mRNA | BC108661.1 | - | 1 | 816 | 5240367 |