siRNA Details
| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126092.1 | - | 3, 4 | 846 | ||
| AK128294.1 | - | 1, 2 | 176 | |||
| BX537595.1 | - | 14, 15 | 4338 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADCY10P1| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126092.1 | - | 3, 4 | 846 | ||
| AK128294.1 | - | 1, 2 | 176 | |||
| BX537595.1 | - | 14, 15 | 4338 | |||