siRNA Details
| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126092.1 | - | 4 | 981 | ||
| AK128294.1 | - | 2 | 311 | |||
| BC126399.1 | - | 3 | 361 | |||
| BX537595.1 | - | 15 | 4473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADCY10P1| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126092.1 | - | 4 | 981 | ||
| AK128294.1 | - | 2 | 311 | |||
| BC126399.1 | - | 3 | 361 | |||
| BX537595.1 | - | 15 | 4473 | |||