siRNA Details
| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126092.1 | - | 5 | 1893 | ||
| AK128294.1 | - | 3 | 610 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADCY10P1| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126092.1 | - | 5 | 1893 | ||
| AK128294.1 | - | 3 | 610 | |||