siRNA Details
| Gene Symbol: DKFZP586I1420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002186.1 | 1 | 2198 | |||
| GenBank mRNA | AL050378.1 | - | 1 | 2198 | ||
| BC064599.1 | - | 1 | 1510 | |||
| BC073840.1 | - | 1 | 2382 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DKFZP586I1420| Gene Symbol: DKFZP586I1420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002186.1 | 1 | 2198 | |||
| GenBank mRNA | AL050378.1 | - | 1 | 2198 | ||
| BC064599.1 | - | 1 | 1510 | |||
| BC073840.1 | - | 1 | 2382 | |||