siRNA Details
| Gene Symbol: MTURN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152793.2 | NP_690006.2 | 2 | 399 | ||
| XM_005249652.3 | XP_005249709.1 | 2 | 342 | |||
| GenBank mRNA | AK096533.1 | - | 2 | 342 | ||
| AK294982.1 | - | 2 | 129 | |||
| AL834391.1 | - | 2 | 333 | |||
| AY054121.1 | - | 2 | 195 | |||
| BC062423.1 | - | 2 | 150 | |||
| BI560774.1 | - | 2 | 403 | |||
| DT217816.1 | - | 2 | 334 | |||

MTURN
Genomic Map