siRNA Details
| Gene Symbol: MTURN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152793.2 | NP_690006.2 | 2 | 441 | ||
| XM_005249652.3 | XP_005249709.1 | 2 | 384 | |||
| GenBank mRNA | AK294982.1 | - | 2 | 171 | ||
| AL834391.1 | - | 2 | 375 | |||
| AY054121.1 | - | 2 | 237 | |||
| BC062423.1 | - | 2 | 192 | |||
| BI560774.1 | - | 2 | 445 | |||
| DT217816.1 | - | 2 | 376 | |||

MTURN
Genomic Map