siRNA Details
| Gene Symbol: PRR15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175887.2 | NP_787083.1 | 2 | 762 | ||
| XM_005249653.3 | XP_005249710.1 | 2 | 1565 | |||
| XM_011515198.1 | XP_011513500.1 | 2 | 1560 | |||
| XM_011515199.1 | XP_011513501.1 | 2 | 474 | |||
| XM_017011842.1 | XP_016867331.1 | 2 | 772 | |||
| GenBank mRNA | BC018144.1 | - | 2 | 659 | ||
| BC029131.1 | - | 2 | 762 | 5189352 | ||
| FJ857942.1 | - | 1 | 82 | |||

PRR15
Genomic Map