siRNA Details
| Gene Symbol: SRRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110199.2 | NP_001103669.1 | 15 | 2433 | ||
| NM_001291831.1 | NP_001278760.1 | 16 | 2512 | |||
| GenBank mRNA | AK092590.1 | - | 3 | 851 | ||
| AK094397.1 | - | 1 | 1044 | |||
| BC063788.1 | - | 1 | 772 | |||
| BC096755.1 | - | 4 | 962 | |||

SRRM3
Genomic Map