siRNA Details
| Gene Symbol: TTTY20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001546.1 | 2 | 186 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTTY20| Gene Symbol: TTTY20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001546.1 | 2 | 186 | |||