siRNA Details
| Gene Symbol: TTTY21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001535.1 | 1 | 88 | |||
| Gene Symbol: TTTY21B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003588.1 | 1 | 88 | |||

TTTY21
Genomic Map