siRNA Details
Gene Symbol: HCG27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026791.1 | 2 | 320 | |||
GenBank mRNA | AK097442.1 | - | 2 | 319 | ||
BC041465.2 | - | 1 | 199 | |||
BC104432.1 | - | 2 | 241 | |||
BC104433.1 | - | 2 | 243 | |||
DB027671.1 | - | 2 | 319 |
