siRNA Details
| Gene Symbol: SERHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284334.1 | NP_001271263.1 | 9 | 728 | ||
| NM_014509.4 | NP_055324.2 | 11 | 920 | |||
| NR_104300.1 | 10 | 1599 | ||||
| NR_104301.1 | 10 | 1583 | ||||
| XM_017028738.1 | XP_016884227.1 | 11 | 948 | |||
| XM_017028739.1 | XP_016884228.1 | 10 | 865 | |||
| GenBank mRNA | AK294584.1 | - | 10 | 1543 | ||
| AL450314.1 | - | 11 | 863 | |||
| AL589866.1 | - | 5 | 221 | |||
| AL590118.1 | - | 9 | 569 | |||
| AL590120.1 | - | 11 | 761 | |||
| BC119650.2 | - | 11 | 804 | 40115836 | ||
| BC119651.1 | - | 11 | 804 | 40115837 | ||
| BC127635.1 | - | 11 | 805 | 40115838 | ||
| BC127636.1 | - | 11 | 805 | 40115841 | ||
| CR456455.1 | - | 5 | 227 | |||

SERHL2
Genomic Map