siRNA Details
| Gene Symbol: LINC01265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104631.1 | 3 | 257 | |||
| GenBank mRNA | AA393775.1 | - | 3 | 255 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01265| Gene Symbol: LINC01265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104631.1 | 3 | 257 | |||
| GenBank mRNA | AA393775.1 | - | 3 | 255 | ||