siRNA Details
| Gene Symbol: LINC01265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104631.1 | 4 | 376 | |||
| GenBank mRNA | AA393775.1 | - | 4 | 373 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01265| Gene Symbol: LINC01265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104631.1 | 4 | 376 | |||
| GenBank mRNA | AA393775.1 | - | 4 | 373 | ||