siRNA Details
| Gene Symbol: ZNF503-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038223.1 | 3 | 822 | |||
| NR_038224.1 | 3 | 480 | ||||
| NR_038225.1 | 4 | 591 | ||||
| NR_038226.1 | 4 | 603 | ||||
| GenBank mRNA | AK023445.1 | - | 3 | 815 | ||
| CB997793.1 | - | 3 | 508 | |||
| CN360505.1 | - | 4 | 489 | |||

ZNF503-AS1
Genomic Map