siRNA Details
| Gene Symbol: ASPHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181718.3 | NP_859069.2 | 1, 2 | 1088 | ||
| XM_017023107.1 | XP_016878596.1 | 2, 3 | 1069 | |||
| GenBank mRNA | AF070642.1 | - | 1, 2 | 429 | ||
| BC009033.1 | - | 1, 2 | 815 | |||
| BC029136.1 | - | 1, 2 | 524 | |||
| BC034275.3 | - | 1, 2 | 561 | |||
| BC040933.2 | - | 1, 2 | 600 | |||
| BC085011.1 | - | 1, 2 | 936 | |||
| BC126319.1 | - | 1, 2 | 1056 | 8992035 | ||
| BC126321.1 | - | 1, 2 | 1056 | 8992037 | ||
| BC144152.1 | - | 1, 2 | 1059 | 9052672 | ||

ASPHD1
Genomic Map