siRNA Details
| Gene Symbol: ASPHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181718.3 | NP_859069.2 | 1 | 567 | ||
| GenBank mRNA | BC009033.1 | - | 1 | 294 | ||
| BC034275.3 | - | 1 | 37 | |||
| BC040933.2 | - | 1 | 79 | |||
| BC085011.1 | - | 1 | 415 | |||
| BC126319.1 | - | 1 | 535 | 8992035 | ||
| BC126321.1 | - | 1 | 535 | 8992037 | ||
| BC144152.1 | - | 1 | 535 | 9052672 | ||

ASPHD1
Genomic Map