siRNA Details
| Gene Symbol: TBC1D28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039397.2 | NP_001034486.2 | 4 | 424 | ||
| XM_011523781.2 | XP_011522083.1 | 4 | 424 | |||
| XM_011523782.2 | XP_011522084.1 | 4 | 304 | |||
| GenBank mRNA | AK097563.1 | - | 3 | 712 | ||
| BC112012.1 | - | 2 | 90 | 8327480 | ||
| BC112016.1 | - | 2 | 90 | 8327484 | ||
| DB065322.1 | - | 4 | 424 | |||

TBC1D28
Genomic Map