siRNA Details
| Gene Symbol: LINC00710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015413.1 | 2 | 316 | |||
| NR_024410.1 | 2 | 316 | ||||
| GenBank mRNA | BC033338.1 | - | 2 | 248 | ||
| BC106956.2 | - | 2 | 120 | |||
| DB038073.1 | - | 2 | 316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00710| Gene Symbol: LINC00710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015413.1 | 2 | 316 | |||
| NR_024410.1 | 2 | 316 | ||||
| GenBank mRNA | BC033338.1 | - | 2 | 248 | ||
| BC106956.2 | - | 2 | 120 | |||
| DB038073.1 | - | 2 | 316 | |||