siRNA Details
| Gene Symbol: LINC00710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015413.1 | 3 | 408 | |||
| NR_024410.1 | 3 | 408 | ||||
| NR_024411.1 | 2 | 297 | ||||
| GenBank mRNA | BC033338.1 | - | 3 | 340 | ||
| BC106956.2 | - | 3 | 212 | |||
| BC106957.2 | - | 2 | 101 | |||
| BC113009.1 | - | 2 | 101 | |||

LINC00710
Genomic Map