siRNA Details
Gene Symbol: C11orf86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136485.1 | NP_001129957.1 | 1 | 128 | ||
GenBank mRNA | AK026328.1 | - | 1 | 134 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C11orf86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136485.1 | NP_001129957.1 | 1 | 128 | ||
GenBank mRNA | AK026328.1 | - | 1 | 134 |