siRNA Details
| Gene Symbol: NUDT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243750.1 | NP_001230679.1 | 3 | 375 | ||
| NM_181843.2 | NP_862826.1 | 3 | 375 | |||
| XM_017017474.1 | XP_016872963.1 | 3 | 554 | |||
| GenBank mRNA | AK123561.1 | - | 3 | 359 | ||
| BC018644.1 | - | 3 | 368 | 3956297 | ||
| CB140633.1 | - | 3 | 375 | |||

NUDT8
Genomic Map