siRNA Details
Gene Symbol: LINC00887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024480.1 | 2 | 286 | |||
GenBank mRNA | AK054628.1 | - | 2 | 289 | ||
BC112168.1 | - | 2 | 120 | |||
BC113537.1 | - | 2 | 120 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024480.1 | 2 | 286 | |||
GenBank mRNA | AK054628.1 | - | 2 | 289 | ||
BC112168.1 | - | 2 | 120 | |||
BC113537.1 | - | 2 | 120 |