siRNA Details
| Gene Symbol: KRTAP15-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181623.1 | NP_853654.1 | 1 | 12 | ||
| GenBank mRNA | AB096942.1 | - | 1 | 34 | ||
| BC104925.1 | - | 1 | 61 | 8143928 | ||
| BC104927.1 | - | 1 | 61 | 8143930 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP15-1| Gene Symbol: KRTAP15-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181623.1 | NP_853654.1 | 1 | 12 | ||
| GenBank mRNA | AB096942.1 | - | 1 | 34 | ||
| BC104925.1 | - | 1 | 61 | 8143928 | ||
| BC104927.1 | - | 1 | 61 | 8143930 | ||