siRNA Details
| Gene Symbol: LINC00879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015400.1 | 5 | 530 | |||
| GenBank mRNA | BC038721.1 | - | 5 | 533 | ||
| BC048119.1 | - | 5 | 531 | |||
| BC104171.1 | - | 4 | 365 | |||
| BC115359.1 | - | 5 | 449 | |||
| BC115360.1 | - | 5 | 482 | |||

LINC00879
Genomic Map