siRNA Details
| Gene Symbol: LINC00879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015400.1 | 5, 6 | 651 | |||
| GenBank mRNA | BC038721.1 | - | 5, 6 | 654 | ||
| BC048119.1 | - | 5, 6 | 652 | |||
| BC104171.1 | - | 4, 5 | 486 | |||
| BC115359.1 | - | 5, 6 | 570 | |||
| BC115360.1 | - | 5, 6 | 603 | |||

LINC00879
Genomic Map