siRNA Details
| Gene Symbol: LINC01018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024423.2 | 3 | 434 | |||
| NR_024424.2 | 1 | 448 | ||||
| GenBank mRNA | AK125283.1 | - | 3 | 396 | ||
| BC043253.2 | - | 1 | 132 | |||
| BC106918.2 | - | 1 | 427 | |||
| BC106919.1 | - | 1 | 430 | |||
| BI602592.1 | - | 1 | 452 | |||

LINC01018
Genomic Map