siRNA Details
| Gene Symbol: LOC255187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122044.1 | 2, 3 | 463 | |||
| GenBank mRNA | AA443842.1 | - | 1, 2 | 347 | ||
| BX106527.1 | - | 2, 3 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC255187| Gene Symbol: LOC255187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122044.1 | 2, 3 | 463 | |||
| GenBank mRNA | AA443842.1 | - | 1, 2 | 347 | ||
| BX106527.1 | - | 2, 3 | 325 | |||