siRNA Details
| Gene Symbol: EPGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270991.1 | NP_001257920.1 | 2, 3 | 171 | ||
| NM_001270993.1 | NP_001257922.1 | 2, 3 | 171 | |||
| XM_005265666.4 | XP_005265723.1 | 2, 3 | 1070 | |||
| XM_011531817.2 | XP_011530119.1 | 2, 3 | 1056 | |||
| GenBank mRNA | AY358920.1 | - | 2, 3 | 154 | ||
| CB150835.1 | - | 2, 3 | 171 | |||
| DQ235265.1 | - | 2, 3 | 107 | |||
| DQ235268.1 | - | 2, 3 | 107 | |||

EPGN
Genomic Map