siRNA Details
Gene Symbol: LINC01619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046159.2 | 4 | 371 | |||
NR_046160.2 | 4 | 533 | ||||
NR_132339.1 | 3 | 458 | ||||
NR_132340.1 | 3 | 458 | ||||
NR_132341.1 | 3 | 458 | ||||
GenBank mRNA | AK055439.1 | - | 3 | 298 | ||
BC029844.1 | - | 4 | 371 | |||
BC107595.1 | - | 3 | 295 | |||
BC118985.1 | - | 4 | 370 | |||
BC122534.1 | - | 3 | 295 | |||
DB121637.1 | - | 3 | 458 |
