siRNA Details
| Gene Symbol: NUDT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318380.1 | NP_001305309.1 | 3 | 282 | ||
| NM_177533.4 | NP_803877.2 | 3 | 282 | |||
| GenBank mRNA | AB087802.1 | - | 3 | 168 | ||
| AK128196.1 | - | 3 | 271 | |||
| AK310449.1 | - | 3 | 259 | |||
| BC041584.1 | - | 3 | 244 | 4810207 | ||
| DA249638.1 | - | 3 | 202 | |||
| DA409636.1 | - | 3 | 265 | |||
| DB096906.1 | - | 3 | 284 | |||

NUDT14
Genomic Map