siRNA Details
| Gene Symbol: CDCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005273733.3 | XP_005273790.1 | 7 | 1016 | ||
| XM_005273734.3 | XP_005273791.1 | 7 | 1016 | |||
| XM_011544743.2 | XP_011543045.1 | 7 | 1016 | |||
| XM_011544747.2 | XP_011543049.1 | 6 | 572 | |||
| XM_011544748.2 | XP_011543050.1 | 7 | 1041 | |||
| XM_017017158.1 | XP_016872647.1 | 7 | 1016 | |||

CDCA5
Genomic Map